accessibility menu, dialog, popup

UserName

CAS RN 978-86-9

IUPAC Name: 4-(triphenylmethyl)phenol
Synonyms: 4-(triphenylmethyl)phenol
Molecular Weight (g/mol): 336.43
Molecular Formula: C25H20O
InChi Key: NIPKXTKKYSKEON-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)