missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethylthio)benzyl Bromide 97.0+%, TCI America™
Supplier: TCI America T23911G
Specifications
| 4-(Trifluoromethylthio)benzyl Bromide | |
| 55°C | |
| 118°C | |
| MFCD00129189 | |
| 3261 | |
| VAYGJBCOCRVROJ-UHFFFAOYSA-N | |
| 1-(bromomethyl)-4-(trifluoromethylsulfanyl)benzene | |
| 2777882 | |
| ≥97.0% (GC) |
| 21101-63-3 | |
| White-Yellow | |
| C8H6BrF3S | |
| 1 g | |
| 4-trifluoromethylthio benzyl bromide, 4-bromomethyl phenyl trifluoromethyl sulfane, 4-trifluoromethylthio benzylbromide, 1-bromomethyl-4-trifluoromethyl sulfanyl benzene, benzene, 1-bromomethyl-4-trifluoromethyl thio, 1-bromomethyl-4-trifluoromethylsulfanyl-benzene, 1-bromomethyl-4-trifluoromethylsulfanyl benzene, acmc-1coe0, 4-trifluoromethylthiobenzyl bromide, 4 trifluoromethylthio benzyl bromide | |
| C1=CC(=CC=C1CBr)SC(F)(F)F | |
| 271.095 | |
| 271.10 | |
| Crystalline Powder |
Chemical Identifiers
| 21101-63-3 | |
| 271.095 | |
| VAYGJBCOCRVROJ-UHFFFAOYSA-N | |
| 2777882 | |
| C1=CC(=CC=C1CBr)SC(F)(F)F |
| C8H6BrF3S | |
| MFCD00129189 | |
| 4-trifluoromethylthio benzyl bromide, 4-bromomethyl phenyl trifluoromethyl sulfane, 4-trifluoromethylthio benzylbromide, 1-bromomethyl-4-trifluoromethyl sulfanyl benzene, benzene, 1-bromomethyl-4-trifluoromethyl thio, 1-bromomethyl-4-trifluoromethylsulfanyl-benzene, 1-bromomethyl-4-trifluoromethylsulfanyl benzene, acmc-1coe0, 4-trifluoromethylthiobenzyl bromide, 4 trifluoromethylthio benzyl bromide | |
| 1-(bromomethyl)-4-(trifluoromethylsulfanyl)benzene |
Safety and Handling
TSCA : No