missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethylthio)benzyl Bromide 97.0+%, TCI America™
$83.89 - $266.56
Chemical Identifiers
| CAS | 21101-63-3 |
|---|---|
| Molecular Formula | C8H6BrF3S |
| Molecular Weight (g/mol) | 271.095 |
| MDL Number | MFCD00129189 |
| InChI Key | VAYGJBCOCRVROJ-UHFFFAOYSA-N |
| Synonym | 4-trifluoromethylthio benzyl bromide, 4-bromomethyl phenyl trifluoromethyl sulfane, 4-trifluoromethylthio benzylbromide, 1-bromomethyl-4-trifluoromethyl sulfanyl benzene, benzene, 1-bromomethyl-4-trifluoromethyl thio, 1-bromomethyl-4-trifluoromethylsulfanyl-benzene, 1-bromomethyl-4-trifluoromethylsulfanyl benzene, acmc-1coe0, 4-trifluoromethylthiobenzyl bromide, 4 trifluoromethylthio benzyl bromide |
| PubChem CID | 2777882 |
| IUPAC Name | 1-(bromomethyl)-4-(trifluoromethylsulfanyl)benzene |
| SMILES | C1=CC(=CC=C1CBr)SC(F)(F)F |
Chemical Identifiers
| 21101-63-3 | |
| 271.095 | |
| VAYGJBCOCRVROJ-UHFFFAOYSA-N | |
| 2777882 | |
| C1=CC(=CC=C1CBr)SC(F)(F)F |
| C8H6BrF3S | |
| MFCD00129189 | |
| 4-trifluoromethylthio benzyl bromide, 4-bromomethyl phenyl trifluoromethyl sulfane, 4-trifluoromethylthio benzylbromide, 1-bromomethyl-4-trifluoromethyl sulfanyl benzene, benzene, 1-bromomethyl-4-trifluoromethyl thio, 1-bromomethyl-4-trifluoromethylsulfanyl-benzene, 1-bromomethyl-4-trifluoromethylsulfanyl benzene, acmc-1coe0, 4-trifluoromethylthiobenzyl bromide, 4 trifluoromethylthio benzyl bromide | |
| 1-(bromomethyl)-4-(trifluoromethylsulfanyl)benzene |
Specifications
| 21101-63-3 | |
| White-Yellow | |
| C8H6BrF3S | |
| 1 g | |
| 4-trifluoromethylthio benzyl bromide, 4-bromomethyl phenyl trifluoromethyl sulfane, 4-trifluoromethylthio benzylbromide, 1-bromomethyl-4-trifluoromethyl sulfanyl benzene, benzene, 1-bromomethyl-4-trifluoromethyl thio, 1-bromomethyl-4-trifluoromethylsulfanyl-benzene, 1-bromomethyl-4-trifluoromethylsulfanyl benzene, acmc-1coe0, 4-trifluoromethylthiobenzyl bromide, 4 trifluoromethylthio benzyl bromide | |
| C1=CC(=CC=C1CBr)SC(F)(F)F | |
| 271.095 | |
| 271.10 | |
| Crystalline Powder |
| 55°C | |
| 118°C | |
| MFCD00129189 | |
| 3261 | |
| VAYGJBCOCRVROJ-UHFFFAOYSA-N | |
| 1-(bromomethyl)-4-(trifluoromethylsulfanyl)benzene | |
| 2777882 | |
| ≥97.0% (GC) | |
| 4-(Trifluoromethylthio)benzyl Bromide |
Safety and Handling
TSCA : No