Learn More
4-(Trifluoromethylthio)aniline, 98%
CAS: 372-16-7 | C7H6F3NS | 193.19 g/mol
Supplier: Thermo Scientific Chemicals L1606706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-(Trifluoromethylthio)aniline | |
| 1.344 | |
| 106°C (222°F) | |
| C7H6F3NS | |
| MFCD00040926 | |
| UN2810 | |
| 4-trifluoromethylthio aniline, 4-trifluoromethyl thio aniline, 4-trifluoromethyl sulfanyl aniline, 4-aminophenyl trifluoromethyl sulfide, 4-aminophenyl trifluoromethyl sulphide, 4-trifluoromethylsulfanyl aniline, benzenamine, 4-trifluoromethyl thio, 4-trifluoromethylthio phenylamine | |
| NC1=CC=C(SC(F)(F)F)C=C1 | |
| 193.19 | |
| 193.19 |
| 372-16-7 | |
| 103°C (11 mmHg) | |
| Unpleasant | |
| 1.528 | |
| 5 g | |
| 2208859 | |
| OHHHTUXVBNGOGI-UHFFFAOYSA-N | |
| 4-[(trifluoromethyl)sulfanyl]aniline | |
| 123054 | |
| 98% |
Chemical Identifiers
| 372-16-7 | |
| 193.19 | |
| OHHHTUXVBNGOGI-UHFFFAOYSA-N | |
| 123054 |
| C7H6F3NS | |
| MFCD00040926 | |
| 4-trifluoromethylthio aniline, 4-trifluoromethyl thio aniline, 4-trifluoromethyl sulfanyl aniline, 4-aminophenyl trifluoromethyl sulfide, 4-aminophenyl trifluoromethyl sulphide, 4-trifluoromethylsulfanyl aniline, benzenamine, 4-trifluoromethyl thio, 4-trifluoromethylthio phenylamine | |
| NC1=CC=C(SC(F)(F)F)C=C1 |
Safety and Handling
GHS H Statement
H301-H312-H332-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 206-744-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only