missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylthio)aniline, 98%

Catalog No. p-7046731
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1606703 1 g
AAL1606706 5 g
AAL1606714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1606703 Supplier Thermo Scientific Chemicals Supplier No. L1606703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 372-16-7 | C7H6F3NS | 193.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 372-16-7
Molecular Formula C7H6F3NS
Molecular Weight (g/mol) 193.19
MDL Number MFCD00040926
InChI Key OHHHTUXVBNGOGI-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio aniline, 4-trifluoromethyl thio aniline, 4-trifluoromethyl sulfanyl aniline, 4-aminophenyl trifluoromethyl sulfide, 4-aminophenyl trifluoromethyl sulphide, 4-trifluoromethylsulfanyl aniline, benzenamine, 4-trifluoromethyl thio, 4-trifluoromethylthio phenylamine
PubChem CID 123054
SMILES NC1=CC=C(SC(F)(F)F)C=C1

Specifications

Density 1.344
Boiling Point 103°C (11 mmHg)
Flash Point 106°C (222°F)
Odor Unpleasant
Refractive Index 1.528
Quantity 1 g
UN Number UN2810
Beilstein 2208859
IUPAC Name 4-(trifluoromethylsulfanyl)aniline
Formula Weight 193.19
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethylthio)aniline
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H312-H332-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 206-744-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only