missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethylbenzoic anhydride, 97%

Catalog No. p-7032847
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3448003 1 g
AAH3448006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3448003 Supplier Thermo Scientific Chemicals Supplier No. H3448003

CAS: 25753-16-6 | C16H8F6O3 | 362.227 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25753-16-6
Molecular Formula C16H8F6O3
Molecular Weight (g/mol) 362.227
MDL Number MFCD00671577
InChI Key FNAWJOBKLWLHTA-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzoic anhydride, tfba, 4-trifluoromethylbenzoic anhydride, 4-trifluoromethyl benzoyl 4-trifluoromethyl benzoate, benzoic acid,4-trifluoromethyl-, anhydride with 4-trifluoromethyl benzoic acid, acmc-1cg53, p-trifluoromethylbenzoic acid anhydride, bis 4-trifluoromethylbenzoic acid anhydride, benzoic acid, 4-trifluoromethyl-, anhydride
PubChem CID 2760733
IUPAC Name [4-(trifluoromethyl)benzoyl] 4-(trifluoromethyl)benzoate
SMILES C1=CC(=CC=C1C(=O)OC(=O)C2=CC=C(C=C2)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 127°C to 131°C
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 362.23
Percent Purity 97%
Chemical Name or Material 4-Trifluoromethylbenzoic anhydride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only