accessibility menu, dialog, popup

UserName

CAS RN 25753-16-6

IUPAC Name: 4-(trifluoromethyl)benzoyl 4-(trifluoromethyl)benzoate
Synonyms: 4-(trifluoromethyl)benzoyl 4-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 362.23
Molecular Formula: C16H8F6O3
InChi Key: FNAWJOBKLWLHTA-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)C(=O)OC(=O)C1=CC=C(C=C1)C(F)(F)F
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)