missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)thiobenzamide 98.0+%, TCI America™
Supplier: TCI America T27715G
Specifications
| 4-(Trifluoromethyl)thiobenzamide | |
| 137°C | |
| MFCD00051806 | |
| 2811 | |
| IPRFNMJROWWFBH-UHFFFAOYSA-N | |
| 4-(trifluoromethyl)benzenecarbothioamide | |
| 2734823 | |
| ≥98.0% (HPLC) |
| 72505-21-6 | |
| C8H6F3NS | |
| 5 g | |
| 4-trifluoromethyl thiobenzamide, 4-trifluoromethyl benzenecarbothioamide, 4-trifluoromethylthiobenzamide, 4-trifluoromethyl benzene-1-carbothioamide, 4-trifluoromethyl benzothioamide, 4-trifluoromethyl-thiobenzamide, benzenecarbothioamide, 4-trifluoromethyl, 4-trifluoromethyl thiobenzamine | |
| C1=CC(=CC=C1C(=S)N)C(F)(F)F | |
| 205.198 | |
| 205.20 | |
| Crystalline Powder |
Chemical Identifiers
| 72505-21-6 | |
| 205.198 | |
| IPRFNMJROWWFBH-UHFFFAOYSA-N | |
| 2734823 | |
| C1=CC(=CC=C1C(=S)N)C(F)(F)F |
| C8H6F3NS | |
| MFCD00051806 | |
| 4-trifluoromethyl thiobenzamide, 4-trifluoromethyl benzenecarbothioamide, 4-trifluoromethylthiobenzamide, 4-trifluoromethyl benzene-1-carbothioamide, 4-trifluoromethyl benzothioamide, 4-trifluoromethyl-thiobenzamide, benzenecarbothioamide, 4-trifluoromethyl, 4-trifluoromethyl thiobenzamine | |
| 4-(trifluoromethyl)benzenecarbothioamide |
Safety and Handling
TSCA : No