missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)thiobenzamide 98.0+%, TCI America™
$74.56 - $294.52
Chemical Identifiers
| CAS | 72505-21-6 |
|---|---|
| Molecular Formula | C8H6F3NS |
| Molecular Weight (g/mol) | 205.198 |
| MDL Number | MFCD00051806 |
| InChI Key | IPRFNMJROWWFBH-UHFFFAOYSA-N |
| Synonym | 4-trifluoromethyl thiobenzamide, 4-trifluoromethyl benzenecarbothioamide, 4-trifluoromethylthiobenzamide, 4-trifluoromethyl benzene-1-carbothioamide, 4-trifluoromethyl benzothioamide, 4-trifluoromethyl-thiobenzamide, benzenecarbothioamide, 4-trifluoromethyl, 4-trifluoromethyl thiobenzamine |
| PubChem CID | 2734823 |
| IUPAC Name | 4-(trifluoromethyl)benzenecarbothioamide |
| SMILES | C1=CC(=CC=C1C(=S)N)C(F)(F)F |
Chemical Identifiers
| 72505-21-6 | |
| 205.198 | |
| IPRFNMJROWWFBH-UHFFFAOYSA-N | |
| 2734823 | |
| C1=CC(=CC=C1C(=S)N)C(F)(F)F |
| C8H6F3NS | |
| MFCD00051806 | |
| 4-trifluoromethyl thiobenzamide, 4-trifluoromethyl benzenecarbothioamide, 4-trifluoromethylthiobenzamide, 4-trifluoromethyl benzene-1-carbothioamide, 4-trifluoromethyl benzothioamide, 4-trifluoromethyl-thiobenzamide, benzenecarbothioamide, 4-trifluoromethyl, 4-trifluoromethyl thiobenzamine | |
| 4-(trifluoromethyl)benzenecarbothioamide |
Specifications
| 72505-21-6 | |
| C8H6F3NS | |
| 5 g | |
| 4-trifluoromethyl thiobenzamide, 4-trifluoromethyl benzenecarbothioamide, 4-trifluoromethylthiobenzamide, 4-trifluoromethyl benzene-1-carbothioamide, 4-trifluoromethyl benzothioamide, 4-trifluoromethyl-thiobenzamide, benzenecarbothioamide, 4-trifluoromethyl, 4-trifluoromethyl thiobenzamine | |
| C1=CC(=CC=C1C(=S)N)C(F)(F)F | |
| 205.198 | |
| 205.20 | |
| Crystalline Powder |
| 137°C | |
| MFCD00051806 | |
| 2811 | |
| IPRFNMJROWWFBH-UHFFFAOYSA-N | |
| 4-(trifluoromethyl)benzenecarbothioamide | |
| 2734823 | |
| ≥98.0% (HPLC) | |
| 4-(Trifluoromethyl)thiobenzamide |
Safety and Handling
TSCA : No