missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethyl-o-phenylenediamine, 98%

CAS: 368-71-8 | C7H7F3N2 | 176.142 g/mol

Supplier:  Thermo Scientific Chemicals A1274906

 View more versions of this product

Catalog No. AAA1274906


Only null left
Explore available promotions

Description

Description

3,4-Diaminobenzotrifluoride is commonly used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Diaminobenzotrifluoride is commonly used as pharmaceutical intermediate.

Solubility
Miscible with water.

Notes
Stable at room temperature in closed containers under normal storage and handling conditions. Incompatible to oxidizing agents. Store in a cool, dry place.
Specifications

Specifications

4-Trifluoromethyl-o-phenylenediamine
58°C to 62°C
MFCD00042456
UN2811
3,4-diaminobenzotrifluoride, 4-trifluoromethyl benzene-1,2-diamine, 4-trifluoromethyl-1,2-phenylenediamine, 4-trifluoromethyl-o-phenylenediamine, 1,2-benzenediamine, 4-trifluoromethyl, 4-trifluoromethyl-1,2-benzenediamine, 3,4-diamino-benzotrifluoride, 3,4-diamino benzotrifluoride
RQWJHUJJBYMJMN-UHFFFAOYSA-N
4-(trifluoromethyl)benzene-1,2-diamine
520822
98%
368-71-8
C7H7F3N2
5 g
909258
Miscible with water.
C1=CC(=C(C=C1C(F)(F)F)N)N
176.142
176.14

Chemical Identifiers

368-71-8
176.142
RQWJHUJJBYMJMN-UHFFFAOYSA-N
520822
C1=CC(=C(C=C1C(F)(F)F)N)N
C7H7F3N2
MFCD00042456
3,4-diaminobenzotrifluoride, 4-trifluoromethyl benzene-1,2-diamine, 4-trifluoromethyl-1,2-phenylenediamine, 4-trifluoromethyl-o-phenylenediamine, 1,2-benzenediamine, 4-trifluoromethyl, 4-trifluoromethyl-1,2-benzenediamine, 3,4-diamino-benzotrifluoride, 3,4-diamino benzotrifluoride
4-(trifluoromethyl)benzene-1,2-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only