missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethyl-o-phenylenediamine, 98%

Catalog No. p-7020702
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1274903 1 g
AAA1274906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1274903 Supplier Thermo Scientific Chemicals Supplier No. A1274903

CAS: 368-71-8 | C7H7F3N2 | 176.142 g/mol

3,4-Diaminobenzotrifluoride is commonly used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Diaminobenzotrifluoride is commonly used as pharmaceutical intermediate.

Solubility
Miscible with water.

Notes
Stable at room temperature in closed containers under normal storage and handling conditions. Incompatible to oxidizing agents. Store in a cool, dry place.

Chemical Identifiers

CAS 368-71-8
Molecular Formula C7H7F3N2
Molecular Weight (g/mol) 176.142
MDL Number MFCD00042456
InChI Key RQWJHUJJBYMJMN-UHFFFAOYSA-N
Synonym 3,4-diaminobenzotrifluoride, 4-trifluoromethyl benzene-1,2-diamine, 4-trifluoromethyl-1,2-phenylenediamine, 4-trifluoromethyl-o-phenylenediamine, 1,2-benzenediamine, 4-trifluoromethyl, 4-trifluoromethyl-1,2-benzenediamine, 3,4-diamino-benzotrifluoride, 3,4-diamino benzotrifluoride
PubChem CID 520822
IUPAC Name 4-(trifluoromethyl)benzene-1,2-diamine
SMILES C1=CC(=C(C=C1C(F)(F)F)N)N

Specifications

Melting Point 58°C to 62°C
Quantity 1 g
UN Number UN2811
Beilstein 909258
Solubility Information Miscible with water.
Formula Weight 176.14
Percent Purity 98%
Chemical Name or Material 4-Trifluoromethyl-o-phenylenediamine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only