missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)mandelic acid, 98%

Catalog No. AAB2226403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2226403 1 g
AAB2226406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2226403 Supplier Thermo Scientific Chemicals Supplier No. B2226403

CAS: 395-35-7 | C9H7F3O3 | 220.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 395-35-7
Molecular Formula C9H7F3O3
Molecular Weight (g/mol) 220.147
MDL Number MFCD00009724
InChI Key SDGXYUQKJPFLDG-UHFFFAOYSA-N
Synonym 4-trifluoromethyl mandelic acid, 2-hydroxy-2-4-trifluoromethyl phenyl acetic acid, 4-trifluoromethylmandelic acid, hydroxy 4-trifluoromethyl phenyl acetic acid, 4-trifluoromethyl phenylglycolic acid, 2-4-trifluoromethyl phenyl-2-hydroxyacetic acid, p-trifluoromethylmandelic acid
PubChem CID 164593
IUPAC Name 2-hydroxy-2-[4-(trifluoromethyl)phenyl]acetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)O)C(F)(F)F

Specifications

Melting Point 129°C to 131°C
Quantity 1 g
Beilstein 2979949
Formula Weight 220.15
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)mandelic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-900-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only