CAS RN 395-35-7
IUPAC Name:
2-hydroxy-2-[4-(trifluoromethyl)phenyl]acetic acid
Synonyms:
hydroxy[4-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol):
220.15
Molecular Formula:
C9H7F3O3
InChi Key:
SDGXYUQKJPFLDG-UHFFFAOYNA-N
SMILES:
OC(C(O)=O)C1=CC=C(C=C1)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
4-(Trifluoromethyl)mandelic acid, 98%
4-(Trifluoromethyl)mandelic acid, 98%
CAS: 395-35-7 Molecular Formula: C9H7F3O3 Molecular Weight (g/mol): 220.147 MDL Number: MFCD00009724 InChI Key: SDGXYUQKJPFLDG-UHFFFAOYSA-N Synonym: 4-trifluoromethyl mandelic acid,2-hydroxy-2-4-trifluoromethyl phenyl acetic acid,4-trifluoromethylmandelic acid,hydroxy 4-trifluoromethyl phenyl acetic acid,4-trifluoromethyl phenylglycolic acid,2-4-trifluoromethyl phenyl-2-hydroxyacetic acid,p-trifluoromethylmandelic acid PubChem CID: 164593 IUPAC Name: 2-hydroxy-2-[4-(trifluoromethyl)phenyl]acetic acid SMILES: C1=CC(=CC=C1C(C(=O)O)O)C(F)(F)F