missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Trifluoromethyl-L-phenylalanine, 95%
CAS: 114926-38-4 | C10H10F3NO2 | 233.19 g/mol
$156.23 - $2206.63
Chemical Identifiers
| CAS | 114926-38-4 |
|---|---|
| Molecular Formula | C10H10F3NO2 |
| Molecular Weight (g/mol) | 233.19 |
| MDL Number | MFCD00044946 |
| InChI Key | CRFFPDBJLGAGQL-QMMMGPOBSA-N |
| Synonym | 4-trifluoromethyl-l-phenylalanine, s-2-amino-3-4-trifluoromethyl phenyl propanoic acid, 2s-2-amino-3-4-trifluoromethyl phenyl propanoic acid, l-4-trifluoromethylphenylalanine, l-4-trifluoromethylphe, l-phenylalanine, 4-trifluoromethyl, 4-trifluromethyl-l-phenylalanine hydrochloride, s-2-amino-3-4-trifluoromethyl phenyl propionic acid, h-phe 4-cf3-oh |
| PubChem CID | 2761501 |
| IUPAC Name | (2S)-2-amino-3-[4-(trifluoromethyl)phenyl]propanoic acid |
| SMILES | C1=CC(=CC=C1CC(C(=O)O)N)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH52170MD
|
Thermo Scientific Chemicals
H52170MD |
250 mg |
Each for $156.23
|
|
|||||
|
AAH5217003
|
Thermo Scientific Chemicals
H5217003 |
1 g |
Each for $534.89
|
|
|||||
|
AAH5217006
|
Thermo Scientific Chemicals
H5217006 |
5 g |
Each for $2,206.63
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications4-fluoro-l-phenylalanine (fF), or 4-trifluoromethyl-l-phenylalanine were introduced inside the chain of peptides to enhances their antimicrobial activity..
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 114926-38-4 | |
| 233.19 | |
| CRFFPDBJLGAGQL-QMMMGPOBSA-N | |
| 2761501 | |
| C1=CC(=CC=C1CC(C(=O)O)N)C(F)(F)F |
| C10H10F3NO2 | |
| MFCD00044946 | |
| 4-trifluoromethyl-l-phenylalanine, s-2-amino-3-4-trifluoromethyl phenyl propanoic acid, 2s-2-amino-3-4-trifluoromethyl phenyl propanoic acid, l-4-trifluoromethylphenylalanine, l-4-trifluoromethylphe, l-phenylalanine, 4-trifluoromethyl, 4-trifluromethyl-l-phenylalanine hydrochloride, s-2-amino-3-4-trifluoromethyl phenyl propionic acid, h-phe 4-cf3-oh | |
| (2S)-2-amino-3-[4-(trifluoromethyl)phenyl]propanoic acid |
Specifications
| 114926-38-4 | |
| MFCD00044946 | |
| 4-trifluoromethyl-l-phenylalanine, s-2-amino-3-4-trifluoromethyl phenyl propanoic acid, 2s-2-amino-3-4-trifluoromethyl phenyl propanoic acid, l-4-trifluoromethylphenylalanine, l-4-trifluoromethylphe, l-phenylalanine, 4-trifluoromethyl, 4-trifluromethyl-l-phenylalanine hydrochloride, s-2-amino-3-4-trifluoromethyl phenyl propionic acid, h-phe 4-cf3-oh | |
| CRFFPDBJLGAGQL-QMMMGPOBSA-N | |
| (2S)-2-amino-3-[4-(trifluoromethyl)phenyl]propanoic acid | |
| 233.19 | |
| 233.19 | |
| 4-Trifluoromethyl-L-phenylalanine |
| C10H10F3NO2 | |
| 5747857 | |
| Slightly soluble in water. | |
| C1=CC(=CC=C1CC(C(=O)O)N)C(F)(F)F | |
| 250 mg | |
| 2761501 | |
| 95% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only