missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethyl-L-phenylalanine, 95%

Catalog No. aah5217003
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5217003 1 g
AAH52170MD 250 mg
AAH5217006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5217003 Supplier Thermo Scientific Chemicals Supplier No. H5217003
Only null left

CAS: 114926-38-4 | C10H10F3NO2 | 233.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-fluoro-l-phenylalanine (fF), or 4-trifluoromethyl-l-phenylalanine were introduced inside the chain of peptides to enhances their antimicrobial activity..

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 114926-38-4
Molecular Formula C10H10F3NO2
Molecular Weight (g/mol) 233.19
MDL Number MFCD00044946
InChI Key CRFFPDBJLGAGQL-QMMMGPOBSA-N
Synonym 4-trifluoromethyl-l-phenylalanine, s-2-amino-3-4-trifluoromethyl phenyl propanoic acid, 2s-2-amino-3-4-trifluoromethyl phenyl propanoic acid, l-4-trifluoromethylphenylalanine, l-4-trifluoromethylphe, l-phenylalanine, 4-trifluoromethyl, 4-trifluromethyl-l-phenylalanine hydrochloride, s-2-amino-3-4-trifluoromethyl phenyl propionic acid, h-phe 4-cf3-oh
PubChem CID 2761501
IUPAC Name (2S)-2-amino-3-[4-(trifluoromethyl)phenyl]propanoic acid
SMILES C1=CC(=CC=C1CC(C(=O)O)N)C(F)(F)F

Specifications

Beilstein 5747857
Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 233.19
Percent Purity 95%
Chemical Name or Material 4-Trifluoromethyl-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only