missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzylamine, 97%

CAS: 3300-51-4 | C8H8F3N | 175.15 g/mol

$180.05 - $653.16

Chemical Identifiers

CAS 3300-51-4
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.15
MDL Number MFCD00010220
InChI Key PRDBLLIPPDOICK-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzylamine, 4-trifluoromethylbenzylamine, 4-trifluoromethyl phenyl methanamine, 1-4-trifluoromethyl phenyl methanamine, 4-trifluoromethyl benzyl amine, p-trifluoromethylbenzyl amine, 4-trifluoromethyl-benzylamine, p-trifluoromethyl benzylamine, benzenemethanamine, 4-trifluoromethyl
PubChem CID 76804
SMILES NCC1=CC=C(C=C1)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2274506
View Documents
Thermo Scientific Chemicals
B2274506
5 g
Each for $180.05
Only null left
 
AAB2274514
View Documents
Thermo Scientific Chemicals
B2274514
25 g
Each for $653.16
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3300-51-4
175.15
PRDBLLIPPDOICK-UHFFFAOYSA-N
76804
C8H8F3N
MFCD00010220
4-trifluoromethyl benzylamine, 4-trifluoromethylbenzylamine, 4-trifluoromethyl phenyl methanamine, 1-4-trifluoromethyl phenyl methanamine, 4-trifluoromethyl benzyl amine, p-trifluoromethylbenzyl amine, 4-trifluoromethyl-benzylamine, p-trifluoromethyl benzylamine, benzenemethanamine, 4-trifluoromethyl
NCC1=CC=C(C=C1)C(F)(F)F

Specifications

3300-51-4
80°C to 82°C (15 mmHg)
C8H8F3N
MFCD00010220
UN2735
Air Sensitive
PRDBLLIPPDOICK-UHFFFAOYSA-N
[4-(trifluoromethyl)phenyl]methanamine
76804
98%
1.229
75°C (167°F)
1.464
5 g
2640698
4-trifluoromethyl benzylamine, 4-trifluoromethylbenzylamine, 4-trifluoromethyl phenyl methanamine, 1-4-trifluoromethyl phenyl methanamine, 4-trifluoromethyl benzyl amine, p-trifluoromethylbenzyl amine, 4-trifluoromethyl-benzylamine, p-trifluoromethyl benzylamine, benzenemethanamine, 4-trifluoromethyl
NCC1=CC=C(C=C1)C(F)(F)F
175.15
175.15
4-(Trifluoromethyl)benzylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 221-971-9

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only