missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzylamine, 97%

Catalog No. AAB2274506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2274506 5 g
AAB2274514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2274506 Supplier Thermo Scientific Chemicals Supplier No. B2274506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3300-51-4 | C8H8F3N | 175.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3300-51-4
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.15
MDL Number MFCD00010220
InChI Key PRDBLLIPPDOICK-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzylamine, 4-trifluoromethylbenzylamine, 4-trifluoromethyl phenyl methanamine, 1-4-trifluoromethyl phenyl methanamine, 4-trifluoromethyl benzyl amine, p-trifluoromethylbenzyl amine, 4-trifluoromethyl-benzylamine, p-trifluoromethyl benzylamine, benzenemethanamine, 4-trifluoromethyl
PubChem CID 76804
SMILES NCC1=CC=C(C=C1)C(F)(F)F

Specifications

Density 1.229
Boiling Point 80°C to 82°C (15 mmHg)
Flash Point 75°C (167°F)
Refractive Index 1.464
Quantity 5 g
UN Number UN2735
Beilstein 2640698
Sensitivity Air Sensitive
IUPAC Name [4-(trifluoromethyl)phenyl]methanamine
Formula Weight 175.15
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)benzylamine
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 221-971-9
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only