missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)benzyl Alcohol 96.0+%, TCI America™
$72.70 - $212.51
Chemical Identifiers
| CAS | 349-95-1 |
|---|---|
| Molecular Formula | C8H7F3O |
| Molecular Weight (g/mol) | 176.138 |
| MDL Number | MFCD00004661 |
| InChI Key | MOOUWXDQAUXZRG-UHFFFAOYSA-N |
| Synonym | 4-trifluoromethyl benzyl alcohol, 4-trifluoromethyl phenyl methanol, p-trifluoromethylbenzyl alcohol, 4-trifluoromethyl benzene methanol, 4-trifluoromethyl benzylic alcohol, benzenemethanol, 4-trifluoromethyl, 4-trifluoromethylbenzyl alcohol, ccris 5117, 4-trifluoromethyl benzylalcohol |
| PubChem CID | 67684 |
| IUPAC Name | [4-(trifluoromethyl)phenyl]methanol |
| SMILES | C1=CC(=CC=C1CO)C(F)(F)F |
Chemical Identifiers
| 349-95-1 | |
| 176.138 | |
| MOOUWXDQAUXZRG-UHFFFAOYSA-N | |
| 67684 | |
| C1=CC(=CC=C1CO)C(F)(F)F |
| C8H7F3O | |
| MFCD00004661 | |
| 4-trifluoromethyl benzyl alcohol, 4-trifluoromethyl phenyl methanol, p-trifluoromethylbenzyl alcohol, 4-trifluoromethyl benzene methanol, 4-trifluoromethyl benzylic alcohol, benzenemethanol, 4-trifluoromethyl, 4-trifluoromethylbenzyl alcohol, ccris 5117, 4-trifluoromethyl benzylalcohol | |
| [4-(trifluoromethyl)phenyl]methanol |
Specifications
| 349-95-1 | |
| 80°C | |
| MFCD00004661 | |
| 4-trifluoromethyl benzyl alcohol, 4-trifluoromethyl phenyl methanol, p-trifluoromethylbenzyl alcohol, 4-trifluoromethyl benzene methanol, 4-trifluoromethyl benzylic alcohol, benzenemethanol, 4-trifluoromethyl, 4-trifluoromethylbenzyl alcohol, ccris 5117, 4-trifluoromethyl benzylalcohol | |
| C1=CC(=CC=C1CO)C(F)(F)F | |
| 176.138 | |
| 176.14 | |
| Crystalline Lumps |
| White-Yellow | |
| C8H7F3O | |
| 5 g | |
| MOOUWXDQAUXZRG-UHFFFAOYSA-N | |
| [4-(trifluoromethyl)phenyl]methanol | |
| 67684 | |
| ≥96.0% (GC) | |
| 4-(Trifluoromethyl)benzyl Alcohol |
Safety and Handling
TSCA : No