missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzyl alcohol, 98%

Catalog No. AAA1050806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1050806 5 g
AAA1050814 25 g
AAA1050822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1050806 Supplier Thermo Scientific Chemicals Supplier No. A1050806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 349-95-1 | C8H7F3O | 176.138 g/mol

4-(Trifluoromethyl)benzyl alcohol, is used as a pharmaceutical intermediate, it is also used to predict the NMR spectrum. 4-(Trifluoromethyl)benzyl alcohol is employed as a reagent in, for example, kinetic studies of phosphonoformate prodrugs and aquachromium(IV).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzyl alcohol, is used as a pharmaceutical intermediate, it is also used to predict the NMR spectrum. 4-(Trifluoromethyl)benzyl alcohol is employed as a reagent in, for example, kinetic studies of phosphonoformate prodrugs and aquachromium(IV).

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Keep away from oxidizing agents,reducing agents, acid chlorides, acid anhydrides.

Chemical Identifiers

CAS 349-95-1
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.138
MDL Number MFCD00004661
InChI Key MOOUWXDQAUXZRG-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzyl alcohol, 4-trifluoromethyl phenyl methanol, p-trifluoromethylbenzyl alcohol, 4-trifluoromethyl benzene methanol, 4-trifluoromethyl benzylic alcohol, benzenemethanol, 4-trifluoromethyl, 4-trifluoromethylbenzyl alcohol, ccris 5117, 4-trifluoromethyl benzylalcohol
PubChem CID 67684
IUPAC Name [4-(trifluoromethyl)phenyl]methanol
SMILES C1=CC(=CC=C1CO)C(F)(F)F

Specifications

Melting Point 18°C to 20°C
Density 1.288
Boiling Point 78°C to 80°C (4 mmHg)
Flash Point 100°C (212°F)
Refractive Index 1.459
Quantity 5 g
Beilstein 1450174
Solubility Information Soluble in water.
Formula Weight 176.14
Percent Purity 98%
Chemical Name or Material 4- (Trifluoromethyl)benzyl alcohol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-494-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only