missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzenethiol 98.0+%, TCI America™

Catalog No. p-7137505
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29721G 1 g
T29725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29721G Supplier TCI America Supplier No. T29721G
Only null left

Chemical Identifiers

CAS 825-83-2
Molecular Formula C7H5F3S
Molecular Weight (g/mol) 178.172
MDL Number MFCD00042320
InChI Key WCMLRSZJUIKVCW-UHFFFAOYSA-N
Synonym 4-trifluoromethyl thiophenol, 4-trifluoromethyl benzenethiol, 4-trifluoromethyl benzene-1-thiol, 4-trifluoromethylthiophenol, 4-mercaptobenzotrifluoride, benzenethiol, 4-trifluoromethyl, 4-trifluoromethyl t, acmc-20amtl, pubchem6864
PubChem CID 136653
IUPAC Name 4-(trifluoromethyl)benzenethiol
SMILES C1=CC(=CC=C1C(F)(F)F)S

Specifications

Color Colorless
Quantity 1 g
UN Number 2810
Formula Weight 178.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethyl)benzenethiol