accessibility menu, dialog, popup

UserName

CAS RN 825-83-2

IUPAC Name: [4-(trifluoromethyl)phenyl]sulfanide
Synonyms: [4-(trifluoromethyl)phenyl]sulfanide
Molecular Weight (g/mol): 177.16
Molecular Formula: C7H4F3S
InChi Key: WCMLRSZJUIKVCW-UHFFFAOYSA-M
SMILES: FC(F)(F)C1=CC=C([S-])C=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)