missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzenesulfonyl chloride, 98%, Thermo Scientific™

Chemical Identifiers

CAS 2991-42-6
Molecular Formula C7H4ClF3O2S
Molecular Weight (g/mol) 244.61
MDL Number MFCD00042422
InChI Key OZDCZHDOIBUGAJ-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzene-1-sulfonyl chloride, 4-trifluoromethyl benzene sulfonyl chloride, p-trifluoromethylbenzenesulfonyl chloride, 4-trifluoromethyl benzenesulfonylchloride, 4-trifluoromethyl-benzenesulfonyl chloride, 4-trifluoromethyl phenylsulfonyl chloride, p-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzenesulphonyl chloride, benzenesulfonyl chloride, 4-trifluoromethyl
PubChem CID 2777399
IUPAC Name 4-(trifluoromethyl)benzenesulfonyl chloride
SMILES FC(F)(F)C1=CC=C(C=C1)S(Cl)(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC397760010
SDS View Documents
Thermo Scientific Chemicals
397760010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC397760050
SDS View Documents
Thermo Scientific Chemicals
397760050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2991-42-6
244.61
OZDCZHDOIBUGAJ-UHFFFAOYSA-N
2777399
FC(F)(F)C1=CC=C(C=C1)S(Cl)(=O)=O
C7H4ClF3O2S
MFCD00042422
4-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzene-1-sulfonyl chloride, 4-trifluoromethyl benzene sulfonyl chloride, p-trifluoromethylbenzenesulfonyl chloride, 4-trifluoromethyl benzenesulfonylchloride, 4-trifluoromethyl-benzenesulfonyl chloride, 4-trifluoromethyl phenylsulfonyl chloride, p-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzenesulphonyl chloride, benzenesulfonyl chloride, 4-trifluoromethyl
4-(trifluoromethyl)benzenesulfonyl chloride

Specifications

2991-42-6
100.0
Authentic
Glass bottle
CF3OC6H4SO2Cl
1g
OZDCZHDOIBUGAJ-UHFFFAOYSA-N
4-(trifluoromethyl)benzenesulfonyl chloride
2777399
98%
97.5
118°C to 120°C (18.0mmHg)
97.5% min. (GC)
C7H4ClF3O2S
MFCD00042422
4-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzene-1-sulfonyl chloride, 4-trifluoromethyl benzene sulfonyl chloride, p-trifluoromethylbenzenesulfonyl chloride, 4-trifluoromethyl benzenesulfonylchloride, 4-trifluoromethyl-benzenesulfonyl chloride, 4-trifluoromethyl phenylsulfonyl chloride, p-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzenesulphonyl chloride, benzenesulfonyl chloride, 4-trifluoromethyl
FC(F)(F)C1=CC=C(C=C1)S(Cl)(=O)=O
244.61
244.62
4-(Trifluoromethyl)benzenesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only