accessibility menu, dialog, popup

UserName

FFFSOOCl

CAS RN 2991-42-6

IUPAC Name: 4-(trifluoromethyl)benzene-1-sulfonyl chloride
Synonyms: 4-(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 244.61
Molecular Formula: C7H4ClF3O2S
InChi Key: OZDCZHDOIBUGAJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)S(Cl)(=O)=O
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)