missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzenesulfonamide 98.0+%, TCI America™

Catalog No. p-7137449
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
25 g
5 g
Catalog No. Quantity
T28471G 1 g
T28475G 5 g
T284725G 25 g
3 options

Catalog No. T28471G

Supplier: TCI America T28471G

Only null left

Chemical Identifiers

CAS 830-43-3
Molecular Formula C7H6F3NO2S
Molecular Weight (g/mol) 225.185
MDL Number MFCD00159251
InChI Key TVHXQQJDMHKGGK-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzenesulfonamide, 4-trifluoromethyl benzene-1-sulfonamide, benzenesulfonamide, p-trifluoromethyl, 4-trifluoromethylbenzenesulphonamide, p-trifluoromethyl benzenesulfonamide, 4-trifluoromethyl benzenesulphonamide, benzenesulfonamide, 4-trifluoromethyl, p-trifluoromethylbenzenesulfonamide, alpha,alpha,alpha-trifluoro-p-toluenesulphonamide, pubchem11767
PubChem CID 70018
IUPAC Name 4-(trifluoromethyl)benzenesulfonamide
SMILES C1=CC(=CC=C1C(F)(F)F)S(=O)(=O)N

Specifications

Melting Point 181°C
Color White
Quantity 1 g
Formula Weight 225.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethyl)benzenesulfonamide
RTECSNumber DB3200000
TSCA No