missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenylacetonitrile 98.0+%, TCI America™

Catalog No. T18041G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18041G 1 g
T18045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18041G Supplier TCI America Supplier No. T18041G

Chemical Identifiers

CAS 49561-96-8
Molecular Formula C9H6F3NO
Molecular Weight (g/mol) 201.15
MDL Number MFCD00061240
InChI Key LYFCAROXYJTUQF-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenylacetonitrile, 2-4-trifluoromethoxy phenyl acetonitrile, 4-trifluoromethoxybenzyl cyanide, 4-trifluoromethoxy benzyl cyanide, benzeneacetonitrile, 4-trifluoromethoxy, 4-trifluoromethoxy-phenyl-acetonitrile, 4-trifluoromethoxy phenyl acetonitrile, 4-trifluoromethoxybenzylcyanide, 1-4-trifluoromethoxy phenyl acetonitrile
PubChem CID 142676
IUPAC Name 2-[4-(trifluoromethoxy)phenyl]acetonitrile
SMILES FC(F)(F)OC1=CC=C(CC#N)C=C1

Specifications

Boiling Point 96°C
Quantity 1 g
Formula Weight 201.15
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-(Trifluoromethoxy)phenylacetonitrile