accessibility menu, dialog, popup

UserName

CAS RN 49561-96-8

IUPAC Name: 2-[4-(trifluoromethoxy)phenyl]acetonitrile
Synonyms: 2-[4-(trifluoromethoxy)phenyl]acetonitrile
Molecular Weight (g/mol): 201.15
Molecular Formula: C9H6F3NO
InChi Key: LYFCAROXYJTUQF-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(CC#N)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)