missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethoxy)benzenesulfonyl Chloride 98.0+%, TCI America™
Supplier: TCI America T214625G
Specifications
| 4-(Trifluoromethoxy)benzenesulfonyl Chloride | |
| Yellow | |
| C7H4ClF3O3S | |
| 25 g | |
| 4-trifluoromethoxy benzenesulfonyl chloride, 4-trifluoromethoxy benzene-1-sulfonyl chloride, 4-trifluoromethoxy benzenesulphonyl chloride, benzenesulfonyl chloride, 4-trifluoromethoxy, p-trifluoromethoxy benzenesulphonyl chloride, 4-trifluoromethoxyphenyl-sulfonylchloride, 4-trifluoromethoxyphenylsulfonyl chloride, 4-trifluoromethoxy benzensulfonylchloride, 4-trifluoromethoxy-benzenesulfonylchloride, 4-trifluoromethoxy-benzenesulfonyl chloride | |
| C1=CC(=CC=C1OC(F)(F)F)S(=O)(=O)Cl | |
| 260.611 | |
| 260.61 | |
| Liquid |
| 94108-56-2 | |
| 122°C | |
| MFCD00042408 | |
| 3265 | |
| UHCDBMIOLNKDHG-UHFFFAOYSA-N | |
| 4-(trifluoromethoxy)benzenesulfonyl chloride | |
| 523102 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 94108-56-2 | |
| 260.611 | |
| UHCDBMIOLNKDHG-UHFFFAOYSA-N | |
| 523102 | |
| C1=CC(=CC=C1OC(F)(F)F)S(=O)(=O)Cl |
| C7H4ClF3O3S | |
| MFCD00042408 | |
| 4-trifluoromethoxy benzenesulfonyl chloride, 4-trifluoromethoxy benzene-1-sulfonyl chloride, 4-trifluoromethoxy benzenesulphonyl chloride, benzenesulfonyl chloride, 4-trifluoromethoxy, p-trifluoromethoxy benzenesulphonyl chloride, 4-trifluoromethoxyphenyl-sulfonylchloride, 4-trifluoromethoxyphenylsulfonyl chloride, 4-trifluoromethoxy benzensulfonylchloride, 4-trifluoromethoxy-benzenesulfonylchloride, 4-trifluoromethoxy-benzenesulfonyl chloride | |
| 4-(trifluoromethoxy)benzenesulfonyl chloride |
Safety and Handling
TSCA : No