missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7137100
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T21465G 5 g
T214625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T21465G Supplier TCI America Supplier No. T21465G
Only null left

Chemical Identifiers

CAS 94108-56-2
Molecular Formula C7H4ClF3O3S
Molecular Weight (g/mol) 260.611
MDL Number MFCD00042408
InChI Key UHCDBMIOLNKDHG-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzenesulfonyl chloride, 4-trifluoromethoxy benzene-1-sulfonyl chloride, 4-trifluoromethoxy benzenesulphonyl chloride, benzenesulfonyl chloride, 4-trifluoromethoxy, p-trifluoromethoxy benzenesulphonyl chloride, 4-trifluoromethoxyphenyl-sulfonylchloride, 4-trifluoromethoxyphenylsulfonyl chloride, 4-trifluoromethoxy benzensulfonylchloride, 4-trifluoromethoxy-benzenesulfonylchloride, 4-trifluoromethoxy-benzenesulfonyl chloride
PubChem CID 523102
IUPAC Name 4-(trifluoromethoxy)benzenesulfonyl chloride
SMILES C1=CC(=CC=C1OC(F)(F)F)S(=O)(=O)Cl

Specifications

Color Yellow
Boiling Point 122°C
Quantity 5 g
UN Number 3265
Formula Weight 260.61
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)benzenesulfonyl Chloride