missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(trans-4-Propylcyclohexyl)phenol 98.0+%, TCI America™
Supplier: TCI America P1616100G
Specifications
| 4-(trans-4-Propylcyclohexyl)phenol | |
| 138°C | |
| C15H22O | |
| 100 g | |
| AHAZEMSUUYFDMM-UHFFFAOYSA-N | |
| 4-(4-propylcyclohexyl)phenol | |
| 3126231 | |
| ≥98.0% (GC) |
| 81936-33-6 | |
| White-Yellow | |
| MFCD00673751 | |
| 1-Hydroxy-4-(trans-4-propylcyclohexyl)benzene | |
| CCCC1CCC(CC1)C2=CC=C(C=C2)O | |
| 218.34 | |
| 218.34 | |
| Crystalline Powder |
Chemical Identifiers
| 81936-33-6 | |
| 218.34 | |
| AHAZEMSUUYFDMM-UHFFFAOYSA-N | |
| 3126231 | |
| CCCC1CCC(CC1)C2=CC=C(C=C2)O |
| C15H22O | |
| MFCD00673751 | |
| 1-Hydroxy-4-(trans-4-propylcyclohexyl)benzene | |
| 4-(4-propylcyclohexyl)phenol |
Safety and Handling
TSCA : No