missing translation for 'onlineSavingsMsg'
Learn More

4-(trans-4-Butylcyclohexyl)phenol 98.0+%, TCI America™

Catalog No. p-7119705
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B264425G 25 g
B2644100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B264425G Supplier TCI America Supplier No. B264425G
Only null left

Chemical Identifiers

CAS 88581-00-4
Molecular Formula C16H24O
Molecular Weight (g/mol) 232.367
MDL Number MFCD00673752
InChI Key SJXVTMAQPHVBEG-UHFFFAOYSA-N
Synonym 1-(trans-4-Butylcyclohexyl)-4-hydroxybenzene
PubChem CID 19845546
IUPAC Name 4-(4-butylcyclohexyl)phenol
SMILES CCCCC1CCC(CC1)C2=CC=C(C=C2)O

Specifications

Color White
Quantity 25 g
Formula Weight 232.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(trans-4-Butylcyclohexyl)phenol