missing translation for 'onlineSavingsMsg'
Learn More

4-(trans-4-Amylcyclohexyl)benzoic Acid 98.0+%, TCI America™

Catalog No. p-7117981
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25935G 5 g
A259325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25935G Supplier TCI America Supplier No. A25935G
Only null left

Chemical Identifiers

CAS 65355-30-8
Molecular Formula C18H26O2
Molecular Weight (g/mol) 274.404
MDL Number MFCD06208360
InChI Key YXKKMVGGPRVHIL-UHFFFAOYSA-N
Synonym 4-4-pentylcyclohexyl benzoic acid, 4-trans-4-n-pentylcyclohexyl benzoic acid, 4-trans-4-pentylcyclohexyl benzoic acid, 4-1s,4r-4-pentylcyclohexyl benzoic acid, 4-pentyl-cyclohexyl benzoic acid, 4-pentyl cyclohexyl benzoic acid, yxkkmvggprvhil-shtzxodssa-n, 4-4-pentyl-cyclohexyl-benzoic acid, 4-trans-4-amylcyclohexyl benzoic acid
PubChem CID 3104716
IUPAC Name 4-(4-pentylcyclohexyl)benzoic acid
SMILES CCCCCC1CCC(CC1)C2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 181°C
Color White
Quantity 5 g
Formula Weight 274.40
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(trans-4-Amylcyclohexyl)benzoic Acid