missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylthiophenol, 97%

Catalog No. p-7081950
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1214803 1 g
AAA1214806 5 g
AAA1214814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1214803 Supplier Thermo Scientific Chemicals Supplier No. A1214803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2396-68-1 | C10H14S | 166.282 g/mol

4-tert-butylthiophenol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-butylthiophenol is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents.

Chemical Identifiers

CAS 2396-68-1
Molecular Formula C10H14S
Molecular Weight (g/mol) 166.282
MDL Number MFCD00022067
InChI Key GNXBFFHXJDZGEK-UHFFFAOYSA-N
Synonym 4-tert-butylthiophenol, p-tert-butylthiophenol, benzenethiol, p-tert-butyl, p-tert-butylbenzenethiol, p-t-butylthiophenol, p-tert-butylphenyl mercaptan, benzenethiol, 4-1,1-dimethylethyl, benzenethiol, p-tert-butylthio, 4-tert-butyl thiophenol, 4-t-butylthiophenol
PubChem CID 75454
IUPAC Name 4-tert-butylbenzenethiol
SMILES CC(C)(C)C1=CC=C(C=C1)S

Specifications

Melting Point -11°C
Density 0.99
Boiling Point 238°C
Flash Point >110°C (230°F)
Odor Stench
Refractive Index 1.5485
Quantity 1 g
Beilstein 606476
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 166.29
Percent Purity 97%
Chemical Name or Material 4-tert-Butylthiophenol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-255-6
RTECSNumber DC1025000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only