accessibility menu, dialog, popup

UserName

CH3H3CCH3SH

CAS RN 2396-68-1

IUPAC Name: 4-tert-butylbenzene-1-thiol
Synonyms: benzenethiol, p-tert-butyl-
Molecular Weight (g/mol): 166.28
Molecular Formula: C10H14S
InChi Key: GNXBFFHXJDZGEK-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(S)C=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)