missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butylthiophenol, 97%
Description
4-tert-butylthiophenol is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-tert-butylthiophenol is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents.
4-tert-butylthiophenol is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | -11°C |
| Density | 0.99 |
| Boiling Point | 238°C |
| Flash Point | >110°C (230°F) |
| Odor | Stench |
| Refractive Index | 1.5485 |
| Quantity | 5 g |
| Beilstein | 606476 |
| Sensitivity | Air Sensitive |
| Solubility Information | Insoluble in water. |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 219-255-6 |
| RTECSNumber | DC1025000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only