missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylpyrocatechol 98.0+%, TCI America™

Catalog No. p-7118649
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B073925G 25 g
B0739500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B073925G Supplier TCI America Supplier No. B073925G
Only null left

Chemical Identifiers

CAS 98-29-3
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00002201
InChI Key XESZUVZBAMCAEJ-UHFFFAOYSA-N
Synonym 4-tert-butylcatechol, p-tert-butylcatechol, 4-tert-butylpyrocatechol, p-tert-butyl catechol, 4-tert-butyl benzene-1,2-diol, 4-t-butylpyrocatechol, p-tert-butylpyrocatechol, 4-t-butylcatechol, 1,2-benzenediol, 4-1,1-dimethylethyl, 4-tert-butylcatechin
PubChem CID 7381
IUPAC Name 4-tert-butylbenzene-1,2-diol
SMILES CC(C)(C)C1=CC=C(O)C(O)=C1

Specifications

Melting Point 58°C
Boiling Point 285°C
Quantity 25 g
UN Number 2811
Formula Weight 166.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylpyrocatechol
EINECSNumber (3)-0548
RTECSNumber UX1400000
TSCA Yes