accessibility menu, dialog, popup

UserName

CH3H3CCH3OHHO

CAS RN 98-29-3

IUPAC Name: 4-tert-butylbenzene-1,2-diol
Synonyms: 4-tert-butylcatechol
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: XESZUVZBAMCAEJ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(O)C(O)=C1
Boiling Point 
  • (3)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (7)
  • (3)
  • (2)
  • (4)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)