missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenylacetic Acid 98.0+%, TCI America™

Catalog No. p-7123460
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43075G 5 g
B430725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43075G Supplier TCI America Supplier No. B43075G

Chemical Identifiers

CAS 32857-63-9
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00082593
InChI Key RUAYXHSDAMWEDR-UHFFFAOYSA-N
Synonym 4-tert-butylphenylacetic acid, 4-tert-butylphenyl acetic acid, 2-4-tert-butylphenyl acetic acid, 4-t-butylphenylacetic acid, 2-4-tert-butyl phenyl acetic acid, benzeneacetic acid, 4-1,1-dimethylethyl, 4-tert-butyl-phenyl-acetic acid, tb2, acmc-1cq6j, asischem y85510
PubChem CID 118343
IUPAC Name 2-(4-tert-butylphenyl)acetic acid
SMILES CC(C)(C)C1=CC=C(C=C1)CC(=O)O

Specifications

Melting Point 81°C
Color White
Quantity 5 g
Formula Weight 192.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylphenylacetic Acid