accessibility menu, dialog, popup

UserName

CAS RN 32857-63-9

IUPAC Name: 2-(4-tert-butylphenyl)acetate
Synonyms: 2-(4-tert-butylphenyl)acetate
Molecular Weight (g/mol): 191.25
Molecular Formula: C12H15O2
InChi Key: RUAYXHSDAMWEDR-UHFFFAOYSA-M
SMILES: CC(C)(C)C1=CC=C(CC([O-])=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results