missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenyl Glycidyl Ether 98.0+%, TCI America™

Catalog No. B483125G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B483125G 25 g
B4831100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B483125G Supplier TCI America Supplier No. B483125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3101-60-8
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00005136
InChI Key HHRACYLRBOUBKM-UHFFFAOYSA-N
Synonym p-tert-butylphenyl glycidyl ether, 4-tert-butylphenyl glycidyl ether, 4-t-butylphenyl glycidyl ether, 2-4-tert-butylphenoxy methyl oxirane, tert-butylphenol glycidyl ether, tert-butylphenyl glycidyl ether, t-butyl phenyl glycidyl ether, ccris 2638, oxirane, 4-1,1-dimethylethyl phenoxy methyl, 2-4-tert-butyl phenoxy methyl oxirane
PubChem CID 18360
IUPAC Name 2-[(4-tert-butylphenoxy)methyl]oxirane
SMILES CC(C)(C)C1=CC=C(C=C1)OCC2CO2

Specifications

Color Colorless
Boiling Point 294°C
Quantity 25 g
Formula Weight 206.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylphenyl Glycidyl Ether
EINECSNumber (3)-0575&(3)-0594
RTECSNumber TX4250000
TSCA Yes