accessibility menu, dialog, popup

UserName

CAS RN 3101-60-8

IUPAC Name: 2-[(4-tert-butylphenoxy)methyl]oxirane
Synonyms: 2-(4-tert-butylphenoxymethyl)oxirane
Molecular Weight (g/mol): 206.29
Molecular Formula: C13H18O2
InChi Key: HHRACYLRBOUBKM-UHFFFAOYNA-N
SMILES: CC(C)(C)C1=CC=C(OCC2CO2)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)