missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenoxyacetyl chloride, 98%

Catalog No. aaa1940818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1940818 50 g
AAA1940809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1940818 Supplier Thermo Scientific Chemicals Supplier No. A1940818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 90734-55-7 | C12H15ClO2 | 226.70 g/mol

4-tert-Butylphenoxyacetyl chloride is used as a reagent and as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylphenoxyacetyl chloride is used as a reagent and as intermediates.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, alcohol, amine, bases, alkali metals, water, moisture.

Chemical Identifiers

CAS 90734-55-7
Molecular Formula C12H15ClO2
Molecular Weight (g/mol) 226.70
MDL Number MFCD00798568
InChI Key CFTNMBDQWVPSHI-UHFFFAOYSA-N
Synonym 4-tert-butylphenoxyacetyl chloride, 4-tert-butylphenoxy acetyl chloride, 2-4-tert-butylphenoxy acetyl chloride, 2-4-tert-butyl phenoxy acetyl chloride, pubchem14851, 4-tert-butylphenoxyacetyl cloride, para-t-butylphenoxyacetyl chloride, 4-t-butyl-phenoxy acetyl chloride, 4-tert-butyl-phenoxyacetyl chloride
PubChem CID 611195
IUPAC Name 2-(4-tert-butylphenoxy)acetyl chloride
SMILES CC(C)(C)C1=CC=C(OCC(Cl)=O)C=C1

Specifications

Boiling Point 130°C to 132°C (5 mmHg)
Refractive Index 1.5165
Quantity 50 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 226.71
Percent Purity 98%
Chemical Name or Material 4-tert-Butylphenoxyacetyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only