accessibility menu, dialog, popup

UserName

CAS RN 90734-55-7

IUPAC Name: 2-(4-tert-butylphenoxy)acetyl chloride
Synonyms: 2-(4-tert-butylphenoxy)acetyl chloride
Molecular Weight (g/mol): 226.7
Molecular Formula: C12H15ClO2
InChi Key: CFTNMBDQWVPSHI-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(OCC(Cl)=O)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results