missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenoxyacetyl chloride, 98%

CAS: 90734-55-7 | C12H15ClO2 | 226.70 g/mol

Supplier:  Thermo Scientific Chemicals A1940809

 View more versions of this product

Catalog No. AAA1940809


Only null left
Explore available promotions

Description

Description

4-tert-Butylphenoxyacetyl chloride is used as a reagent and as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylphenoxyacetyl chloride is used as a reagent and as intermediates.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, alcohol, amine, bases, alkali metals, water, moisture.
Specifications

Specifications

4-tert-Butylphenoxyacetyl chloride
130°C to 132°C (5 mmHg)
1.5165
10 g
Moisture sensitive
Reacts with water.
CC(C)(C)C1=CC=C(OCC(Cl)=O)C=C1
226.70
226.71
90734-55-7
C12H15ClO2
MFCD00798568
UN3265
4-tert-butylphenoxyacetyl chloride, 4-tert-butylphenoxy acetyl chloride, 2-4-tert-butylphenoxy acetyl chloride, 2-4-tert-butyl phenoxy acetyl chloride, pubchem14851, 4-tert-butylphenoxyacetyl cloride, para-t-butylphenoxyacetyl chloride, 4-t-butyl-phenoxy acetyl chloride, 4-tert-butyl-phenoxyacetyl chloride
CFTNMBDQWVPSHI-UHFFFAOYSA-N
2-(4-tert-butylphenoxy)acetyl chloride
611195
98%

Chemical Identifiers

90734-55-7
226.70
CFTNMBDQWVPSHI-UHFFFAOYSA-N
611195
CC(C)(C)C1=CC=C(OCC(Cl)=O)C=C1
C12H15ClO2
MFCD00798568
4-tert-butylphenoxyacetyl chloride, 4-tert-butylphenoxy acetyl chloride, 2-4-tert-butylphenoxy acetyl chloride, 2-4-tert-butyl phenoxy acetyl chloride, pubchem14851, 4-tert-butylphenoxyacetyl cloride, para-t-butylphenoxyacetyl chloride, 4-t-butyl-phenoxy acetyl chloride, 4-tert-butyl-phenoxyacetyl chloride
2-(4-tert-butylphenoxy)acetyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only