missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcatechol, 99%

Catalog No. AC154351000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154351000 100 g Plastic bottle
AC154355000 500 g Plastic bottle
AC154350025 2.5 kg Plastic bottle
AC154350100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC154351000 Supplier Thermo Scientific Chemicals Supplier No. 154351000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98-29-3 | C10H14O2 | 166.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-29-3
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00002201
InChI Key XESZUVZBAMCAEJ-UHFFFAOYSA-N
Synonym 4-tert-butylcatechol, p-tert-butylcatechol, 4-tert-butylpyrocatechol, p-tert-butyl catechol, 4-tert-butyl benzene-1,2-diol, 4-t-butylpyrocatechol, p-tert-butylpyrocatechol, 4-t-butylcatechol, 1,2-benzenediol, 4-1,1-dimethylethyl, 4-tert-butylcatechin
PubChem CID 7381
IUPAC Name 4-tert-butylbenzene-1,2-diol
SMILES CC(C)(C)C1=CC=C(O)C(O)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 53.0°C to 56.0°C
Density 1.0490g/mL
Boiling Point 285.0°C
Flash Point 129°C
Assay Percent Range 0.7% max. Catechol (GC)
Packaging Plastic bottle
Linear Formula (CH3)3CC6H3(OH)2
Quantity 100 g
Beilstein 06,IV,6014
Fieser 15,54
Specific Gravity 1.049
Solubility Information Solubility in water: 0.2% (25°C). Other solubilities: soluble in hot water,very sol. in alcohol,ethers,esters and ketones,partly sol. in chlorinated and aromatic hydrocarb.,slightly sol. in aliphatics. 240g/100g acetone,80g/100g benzene;60g/100g
Viscosity 90 mm2/s (25°C)
Formula Weight 166.22
Percent Purity 99%
Grade Technical
Chemical Name or Material 4-tert-Butylcatechol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life.
Toxic to aquatic life with long lasting eff
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 202-653-9
RTECSNumber UX1400000
TSCA TSCA

RUO – Research Use Only