Learn More
4-tert-Butylcatechol, 98%
CAS: 98-29-3 | C10H14O2 | 166.22 g/mol
Supplier: Thermo Scientific Chemicals A145990I
Description
4-tert-Butylcatechol is widely utilized as an inhibitor in polymerization of butadiene, styrene, vinyl acetate and other reactive monomers. It plays an important role in the synthesis of tungsten oxide nanoparticles by nonaqueous sol-gel process. It acts as a stabilizer in the manufacturing of polyurethane foam. It is employed as an antioxidant for synthetic rubber, polymers and oil derivatives. It is also utilized as a purification agent for aminoformate catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-tert-Butylcatechol is widely utilized as an inhibitor in polymerization of butadiene, styrene, vinyl acetate and other reactive monomers. It plays an important role in the synthesis of tungsten oxide nanoparticles by nonaqueous sol-gel process. It acts as a stabilizer in the manufacturing of polyurethane foam. It is employed as an antioxidant for synthetic rubber, polymers and oil derivatives. It is also utilized as a purification agent for aminoformate catalysts.
Solubility
Soluble in ether, acetone, alcohol, and methanol. Insoluble in water.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
| 4-tert-Butylcatechol | |
| 53°C to 57°C | |
| 284°C to 286°C | |
| Odorless | |
| MFCD00002201 | |
| UN2923 | |
| Hygroscopic | |
| Soluble in ether,acetone,alcohol,and methanol. Insoluble in water. | |
| CC(C)(C)C1=CC=C(O)C(O)=C1 | |
| 166.22 | |
| 166.22 |
| 98-29-3 | |
| 1.049 | |
| 151°C (303°F) | |
| C10H14O2 | |
| 5000 g | |
| 2043335 | |
| 4-tert-butylcatechol, p-tert-butylcatechol, 4-tert-butylpyrocatechol, p-tert-butyl catechol, 4-tert-butyl benzene-1,2-diol, 4-t-butylpyrocatechol, p-tert-butylpyrocatechol, 4-t-butylcatechol, 1,2-benzenediol, 4-1,1-dimethylethyl, 4-tert-butylcatechin | |
| XESZUVZBAMCAEJ-UHFFFAOYSA-N | |
| 4-tert-butylbenzene-1,2-diol | |
| 7381 | |
| 98% |
Chemical Identifiers
| 98-29-3 | |
| 166.22 | |
| XESZUVZBAMCAEJ-UHFFFAOYSA-N | |
| 7381 | |
| CC(C)(C)C1=CC=C(O)C(O)=C1 |
| C10H14O2 | |
| MFCD00002201 | |
| 4-tert-butylcatechol, p-tert-butylcatechol, 4-tert-butylpyrocatechol, p-tert-butyl catechol, 4-tert-butyl benzene-1,2-diol, 4-t-butylpyrocatechol, p-tert-butylpyrocatechol, 4-t-butylcatechol, 1,2-benzenediol, 4-1,1-dimethylethyl, 4-tert-butylcatechin | |
| 4-tert-butylbenzene-1,2-diol |
Safety and Handling
GHS H Statement
H311-H314-H318-H317
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H302+H312-H314-H317-H335-H350
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 202-653-9
RTECSNumber : UX1400000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only