missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcatechol, 98%

CAS: 98-29-3 | C10H14O2 | 166.22 g/mol

Supplier:  Thermo Scientific Chemicals A145990B

 View more versions of this product

Catalog No. AAA145990B

Only null left
Explore available promotions

Description

Description

4-tert-Butylcatechol is widely utilized as an inhibitor in polymerization of butadiene, styrene, vinyl acetate and other reactive monomers. It plays an important role in the synthesis of tungsten oxide nanoparticles by nonaqueous sol-gel process. It acts as a stabilizer in the manufacturing of polyurethane foam. It is employed as an antioxidant for synthetic rubber, polymers and oil derivatives. It is also utilized as a purification agent for aminoformate catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylcatechol is widely utilized as an inhibitor in polymerization of butadiene, styrene, vinyl acetate and other reactive monomers. It plays an important role in the synthesis of tungsten oxide nanoparticles by nonaqueous sol-gel process. It acts as a stabilizer in the manufacturing of polyurethane foam. It is employed as an antioxidant for synthetic rubber, polymers and oil derivatives. It is also utilized as a purification agent for aminoformate catalysts.

Solubility
Soluble in ether, acetone, alcohol, and methanol. Insoluble in water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

4-tert-Butylcatechol
53°C to 57°C
284°C to 286°C
Odorless
MFCD00002201
UN2923
Hygroscopic
Soluble in ether,acetone,alcohol,and methanol. Insoluble in water.
CC(C)(C)C1=CC=C(O)C(O)=C1
166.22
166.22
98-29-3
1.049
151°C (303°F)
C10H14O2
1000 g
2043335
4-tert-butylcatechol, p-tert-butylcatechol, 4-tert-butylpyrocatechol, p-tert-butyl catechol, 4-tert-butyl benzene-1,2-diol, 4-t-butylpyrocatechol, p-tert-butylpyrocatechol, 4-t-butylcatechol, 1,2-benzenediol, 4-1,1-dimethylethyl, 4-tert-butylcatechin
XESZUVZBAMCAEJ-UHFFFAOYSA-N
4-tert-butylbenzene-1,2-diol
7381
98%

Chemical Identifiers

98-29-3
166.22
XESZUVZBAMCAEJ-UHFFFAOYSA-N
7381
CC(C)(C)C1=CC=C(O)C(O)=C1
C10H14O2
MFCD00002201
4-tert-butylcatechol, p-tert-butylcatechol, 4-tert-butylpyrocatechol, p-tert-butyl catechol, 4-tert-butyl benzene-1,2-diol, 4-t-butylpyrocatechol, p-tert-butylpyrocatechol, 4-t-butylcatechol, 1,2-benzenediol, 4-1,1-dimethylethyl, 4-tert-butylcatechin
4-tert-butylbenzene-1,2-diol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H314-H318-H317
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H302+H312-H314-H317-H335-H350

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 202-653-9

RTECSNumber : UX1400000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only