missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzyl Chloride 89.0+%, TCI America™

Catalog No. B22355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22355G 5 g
B223525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22355G Supplier TCI America Supplier No. B22355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19692-45-6
Molecular Formula C11H15Cl
Molecular Weight (g/mol) 182.69
MDL Number MFCD00000918
InChI Key WAXIFMGAKWIFDQ-UHFFFAOYSA-N
Synonym 4-tert-butylbenzyl chloride, 1-tert-butyl-4-chloromethyl benzene, p-tert-butylbenzyl chloride, 4-tert-butyl benzyl chloride, benzene, 1-chloromethyl-4-1,1-dimethylethyl, 4-tert-butyl benzylchloride, p-t-butylbenzyl chloride, 1-chloromethyl-4-tert-butylbenzene, p-tert-butyl-.alpha.-chlorotoluene
PubChem CID 88198
IUPAC Name 1-tert-butyl-4-(chloromethyl)benzene
SMILES CC(C)(C)C1=CC=C(CCl)C=C1

Specifications

Color Yellow
Quantity 5 g
UN Number 1760
Formula Weight 182.69
Percent Purity ≥89.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylbenzyl Chloride