missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzyl Alcohol 97.0+%, TCI America™

Catalog No. p-7119246
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17615G 5 g
B176125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17615G Supplier TCI America Supplier No. B17615G
Only null left

Chemical Identifiers

CAS 877-65-6
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00004662
InChI Key FVEINXLJOJPHLH-UHFFFAOYSA-N
Synonym 4-tert-butylbenzyl alcohol, 4-tert-butyl phenyl methanol, 4-tert-butylphenyl methanol, 4-t-butylbenzyl alcohol, p-tert-butylbenzyl alcohol, 4-t-butylbenzylalcohol, benzenemethanol, 4-1,1-dimethylethyl, 4-1,1-dimethylethyl benzenemethanol, 4-tert-butylbenzylalcohol, p-t-butylbenzyl alcohol
PubChem CID 13416
IUPAC Name (4-tert-butylphenyl)methanol
SMILES CC(C)(C)C1=CC=C(C=C1)CO

Specifications

Color Yellow
Boiling Point 238°C
Quantity 5 g
Formula Weight 164.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylbenzyl Alcohol