missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzyl alcohol, 98%

Catalog No. AAB2126006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2126006 5 g
AAB2126014 25 g
AAB2126022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2126006 Supplier Thermo Scientific Chemicals Supplier No. B2126006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 877-65-6 | C11H16O | 164.248 g/mol

4-tert-Butylbenzyl alcohol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylbenzyl alcohol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in alcohol and insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Keep away from strong oxidizing agents, acid chlorides, acid anhydrides, strong acids.

Chemical Identifiers

CAS 877-65-6
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00004662
InChI Key FVEINXLJOJPHLH-UHFFFAOYSA-N
Synonym 4-tert-butylbenzyl alcohol, 4-tert-butyl phenyl methanol, 4-tert-butylphenyl methanol, 4-t-butylbenzyl alcohol, p-tert-butylbenzyl alcohol, 4-t-butylbenzylalcohol, benzenemethanol, 4-1,1-dimethylethyl, 4-1,1-dimethylethyl benzenemethanol, 4-tert-butylbenzylalcohol, p-t-butylbenzyl alcohol
PubChem CID 13416
IUPAC Name (4-tert-butylphenyl)methanol
SMILES CC(C)(C)C1=CC=C(C=C1)CO

Specifications

Density 0.928
Boiling Point 139°C to 140°C (20 mmHg)
Flash Point 128°C (262°F)
Refractive Index 1.517
Quantity 5 g
Beilstein 1860665
Solubility Information Soluble in alcohol and insoluble in water.
Formula Weight 164.25
Percent Purity 98%
Chemical Name or Material 4-tert-Butylbenzyl alcohol
EINECSNumber 212-894-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only