missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzonitrile 98.0+%, TCI America™

Catalog No. B15765ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15765ML 5 mL
B157625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15765ML Supplier TCI America Supplier No. B15765ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4210-32-6
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.232
MDL Number MFCD00075840
InChI Key IIZURLNRIMKEDL-UHFFFAOYSA-N
Synonym 4-tert-butyl benzonitrile, 4-t-butylbenzonitrile, benzonitrile, 4-1,1-dimethylethyl, 4-tert-butylcyanobenzene, 4-tert-butyl-benzonitrile, p-t-butylbenzonitrile, acmc-209jnk, p-tert-butylbenzonitrile, benzonitrile, p-tert-butyl, ksc219k6d
PubChem CID 77883
IUPAC Name 4-tert-butylbenzonitrile
SMILES CC(C)(C)C1=CC=C(C=C1)C#N

Specifications

Color Yellow
Boiling Point 112°C
Quantity 5 mL
UN Number 3276
Formula Weight 159.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylbenzonitrile